siRNA Details
| Gene Symbol: Slc44a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145394.3 | NP_663369.2 | 15 | 2073 | ||
| GenBank mRNA | AA450663.1 | - | 2 | 297 | ||
| AK194277.1 | - | 3 | 345 | |||
| BC010552.1 | - | 14 | 1841 | 3582543 | ||
| BC025548.1 | - | 15 | 2071 | 5320841 | ||
| CO813168.1 | - | 2 | 161 | |||

Slc44a3
Genomic Map