siRNA Details
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145416.3 | NP_663391.2 | 9 | 764 | ||
| GenBank mRNA | AK051997.1 | - | 5 | 1544 | ||
| BC021438.1 | - | 9 | 757 | 5039543 | ||
| CO426049.1 | - | 6 | 407 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kri1| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145416.3 | NP_663391.2 | 9 | 764 | ||
| GenBank mRNA | AK051997.1 | - | 5 | 1544 | ||
| BC021438.1 | - | 9 | 757 | 5039543 | ||
| CO426049.1 | - | 6 | 407 | |||