siRNA Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145417.3 | NP_663392.2 | 4, 5 | 938 | ||
| GenBank mRNA | AK145466.1 | - | 4, 5 | 941 | ||
| AK159195.1 | - | 4, 5 | 881 | |||
| AK159445.1 | - | 4, 5 | 870 | |||
| AK159625.1 | - | 4, 5 | 881 | |||
| BC010520.1 | - | 4, 5 | 899 | |||
| BC019200.1 | - | 4, 5 | 909 | 5041005 | ||

Rnpep
Genomic Map