siRNA Details
| Gene Symbol: Wdpcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145425.3 | NP_663400.2 | 12, 13 | 1777 | ||
| GenBank mRNA | AF424701.1 | - | 12, 13 | 1740 | ||
| AK043672.1 | - | 11, 12 | 1664 | |||
| AK044273.1 | - | 1 | 566 | |||
| AK083042.1 | - | 12, 13 | 1778 | |||
| BC056139.1 | - | 3, 4 | 501 | |||
| BC141166.1 | - | 12, 13 | 1741 | 9055778 | ||

Wdpcp
Genomic Map