siRNA Details
| Gene Symbol: Mbip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145442.2 | NP_663417.1 | 5, 6 | 732 | ||
| GenBank mRNA | AK077125.1 | - | 5, 6 | 653 | ||
| AK154318.1 | - | 5, 6 | 661 | |||
| AK171164.1 | - | 5, 6 | 661 | |||
| BC002277.1 | - | 5, 6 | 718 | 3496688 | ||

Mbip
Genomic Map