siRNA Details
| Gene Symbol: BC022687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145450.3 | NP_663425.2 | 1 | 510 | ||
| GenBank mRNA | AK077390.1 | - | 1 | 357 | ||
| AK081297.1 | - | 1 | 511 | |||
| AK161400.1 | - | 1 | 549 | |||
| BC022687.1 | - | 1 | 168 | 4242460 | ||
| BC090976.1 | - | 2 | 329 | 6741627 | ||

BC022687
Genomic Map