siRNA Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145460.2 | NP_663435.2 | 3 | 476 | ||
| GenBank mRNA | AK028649.1 | - | 3 | 386 | ||
| AK031978.1 | - | 2 | 105 | |||
| AK032160.1 | - | 3 | 393 | |||
| AK075924.1 | - | 2 | 103 | |||
| AK139884.1 | - | 3 | 476 | |||
| AK215185.1 | - | 2 | 134 | |||
| BC019806.1 | - | 3 | 254 | 4018387 | ||

Oxnad1
Genomic Map