siRNA Details
| Gene Symbol: Cyp2d34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145474.2 | NP_663449.2 | 2, 3 | 419 | ||
| GenBank mRNA | BC018285.1 | - | 2, 3 | 419 | 4207630 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2d34| Gene Symbol: Cyp2d34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145474.2 | NP_663449.2 | 2, 3 | 419 | ||
| GenBank mRNA | BC018285.1 | - | 2, 3 | 419 | 4207630 | |