siRNA Details
| Gene Symbol: Zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC015291.1 | - | 4 | 318 | 4242025 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp160| Gene Symbol: Zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC015291.1 | - | 4 | 318 | 4242025 | |