siRNA Details
| Gene Symbol: Rhoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145491.2 | NP_663466.2 | 2, 3 | 704 | ||
| GenBank mRNA | AB060650.1 | - | 2, 3 | 522 | ||
| AK180601.1 | - | 2, 3 | 96 | |||
| BC048813.1 | - | 2, 3 | 514 | 2582017 | ||
| BC056363.1 | - | 2, 3 | 704 | 6821501 | ||
| BC057451.1 | - | 2, 3 | 236 | |||

Rhoq
Genomic Map