siRNA Details
| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145497.2 | NP_663472.1 | 3, 4 | 214 | ||
| GenBank mRNA | AK050579.1 | - | 3, 4 | 245 | ||
| AK169962.1 | - | 3, 4 | 188 | |||
| BC016495.1 | - | 3, 4 | 214 | 4221437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmrk1| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145497.2 | NP_663472.1 | 3, 4 | 214 | ||
| GenBank mRNA | AK050579.1 | - | 3, 4 | 245 | ||
| AK169962.1 | - | 3, 4 | 188 | |||
| BC016495.1 | - | 3, 4 | 214 | 4221437 | ||