siRNA Details
| Gene Symbol: Atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145528.3 | NP_663503.1 | 3 | 457 | ||
| GenBank mRNA | AK149672.1 | - | 3 | 416 | ||
| AK154470.1 | - | 3 | 443 | |||
| AK163962.1 | - | 3 | 409 | |||
| AK171206.1 | - | 3 | 443 | |||
| AK171440.1 | - | 3 | 434 | |||
| BC016669.1 | - | 3 | 441 | 2648797 | ||
| BC023673.1 | - | 3 | 407 | 5343236 | ||
| BC023712.1 | - | 3 | 430 | 5346303 | ||
| BC033419.1 | - | 3 | 408 | 5359944 | ||

Atg13
Genomic Map