siRNA Details
| Gene Symbol: Sdcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145535.2 | NP_663510.1 | 5 | 364 | ||
| GenBank mRNA | AK136572.1 | - | 5 | 320 | ||
| AK144452.1 | - | 5 | 329 | |||
| AK160483.1 | - | 5 | 322 | |||
| BC005556.1 | - | 5 | 355 | 3964815 | ||

Sdcbp2
Genomic Map