siRNA Details
| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145542.3 | NP_663517.2 | 1, 2 | 387 | ||
| GenBank mRNA | AB092504.1 | - | 1, 2 | 116 | ||
| AK087454.1 | - | 1, 2 | 345 | |||
| BC018218.1 | - | 1, 2 | 216 | 4007102 | ||
| CJ162898.1 | - | 1, 2 | 400 | |||

Ahcyl1
Genomic Map