siRNA Details
| Gene Symbol: Zfp189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289901.1 | NP_001276830.1 | 4 | 413 | ||
| NM_145547.3 | NP_663522.2 | 3 | 584 | |||
| GenBank mRNA | AK049475.1 | - | 3 | 493 | ||
| AK051265.1 | - | 3 | 543 | |||
| BC021326.1 | - | 3 | 546 | 5065942 | ||
| BC087833.1 | - | 3 | 450 | 5708354 | ||

Zfp189
Genomic Map