siRNA Details
| Gene Symbol: Yipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145550.3 | NP_663525.1 | 4, 5 | 433 | ||
| GenBank mRNA | AK081205.1 | - | 4, 5 | 420 | ||
| AK157536.1 | - | 4, 5 | 420 | |||
| BC009080.1 | - | 4, 5 | 365 | 2655774 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf1| Gene Symbol: Yipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145550.3 | NP_663525.1 | 4, 5 | 433 | ||
| GenBank mRNA | AK081205.1 | - | 4, 5 | 420 | ||
| AK157536.1 | - | 4, 5 | 420 | |||
| BC009080.1 | - | 4, 5 | 365 | 2655774 | ||