siRNA Details
| Gene Symbol: Zfp503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145459.3 | NP_663434.2 | 3 | 1861 | ||
| GenBank mRNA | AK032903.1 | - | 2 | 409 | ||
| AK046771.1 | - | 2 | 875 | |||
| AY281290.1 | - | 2 | 931 | |||
| AY281291.1 | - | 2 | 905 | |||
| BC018484.1 | - | 2 | 847 | 3481880 | ||
| BC058952.1 | - | 2 | 778 | 5715786 | ||
| BC066027.1 | - | 3 | 1861 | 6401144 | ||

Zfp503
Genomic Map