siRNA Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145460.2 | NP_663435.2 | 4 | 540 | ||
| GenBank mRNA | AK028649.1 | - | 4 | 450 | ||
| AK031978.1 | - | 3 | 169 | |||
| AK032160.1 | - | 4 | 457 | |||
| AK075924.1 | - | 3 | 167 | |||
| AK139884.1 | - | 4 | 540 | |||
| AK215115.1 | - | 3 | 198 | |||
| AK215185.1 | - | 3 | 198 | |||
| BC019806.1 | - | 4 | 318 | 4018387 | ||

Oxnad1
Genomic Map