siRNA Details
| Gene Symbol: Zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145483.2 | NP_663458.2 | 4 | 382 | ||
| GenBank mRNA | AK161712.1 | - | 427 | |||
| AK162583.1 | - | 450 | ||||
| AK173216.1 | - | 4 | 413 | |||
| BC015291.1 | - | 4 | 348 | 4242025 | ||

Zfp160
Genomic Map