siRNA Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282012.1 | NP_001268941.1 | 4 | 716 | ||
| NM_001282013.1 | NP_001268942.1 | 6 | 721 | |||
| NM_001282024.1 | NP_001268953.1 | 5 | 815 | |||
| NM_145487.3 | NP_663462.2 | 4 | 744 | |||
| GenBank mRNA | AJ504719.1 | - | 4 | 1513 | ||
| AK132945.1 | - | 4 | 444 | |||
| BC016636.1 | - | 4 | 665 | 2650633 | ||
| BU148736.1 | - | 6 | 678 | |||

Prr3
Genomic Map