siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145505.4 | NP_663480.3 | 4 | 629 | ||
| GenBank mRNA | AK029835.1 | - | 4 | 959 | ||
| AK046518.1 | - | 4 | 621 | |||
| AK054539.1 | - | 4 | 575 | |||
| AK122530.1 | - | 2 | 108 | |||
| AK147386.1 | - | 4 | 610 | |||
| AK172373.1 | - | 4 | 594 | |||
| BC062949.1 | - | 4 | 368 | 6839812 | ||

Fam160b1
Genomic Map