siRNA Details
| Gene Symbol: Rap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145541.5 | NP_663516.1 | 4, 5 | 356 | ||
| GenBank mRNA | AK030897.1 | - | 4, 5 | 306 | ||
| AK152504.1 | - | 4, 5 | 276 | |||
| BC051419.1 | - | 4, 5 | 312 | 3494423 | ||
| BC083128.1 | - | 3, 4 | 208 | 6438722 | ||
| CX206795.1 | - | 4, 5 | 269 | |||

Rap1a
Genomic Map