siRNA Details
| Gene Symbol: Yipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205156.1 | NP_001192085.1 | 7 | 791 | ||
| NM_145550.3 | NP_663525.1 | 8 | 872 | |||
| GenBank mRNA | AK048749.1 | - | 2 | 1643 | ||
| AK081205.1 | - | 8 | 859 | |||
| AK157536.1 | - | 8 | 859 | |||
| AK214559.1 | - | 3 | 341 | |||
| BC009080.1 | - | 8 | 804 | 2655774 | ||

Yipf1
Genomic Map