siRNA Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145460.2 | NP_663435.2 | 8 | 1143 | ||
| GenBank mRNA | AK031978.1 | - | 7 | 772 | ||
| AK032160.1 | - | 8 | 946 | |||
| AK075924.1 | - | 7 | 769 | |||
| AK139884.1 | - | 8 | 1143 | |||
| BC019806.1 | - | 8 | 921 | 4018387 | ||

Oxnad1
Genomic Map