siRNA Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145475.4 | NP_663450.3 | 13 | 2085 | ||
| GenBank mRNA | AB079067.1 | - | 13 | 1592 | ||
| AK042077.1 | - | 13 | 1676 | |||
| AK052269.1 | - | 13 | 1851 | |||
| AK129416.1 | - | 10 | 1227 | |||
| AK161801.1 | - | 923 | ||||
| AK169528.1 | - | 13 | 1670 | |||
| AK197145.1 | - | 2 | 201 | |||
| BC094253.1 | - | 13 | 2084 | 6400872 | ||

Cerk
Genomic Map