siRNA Details
| Gene Symbol: Fam160b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145505.4 | NP_663480.3 | 12 | 1903 | ||
| GenBank mRNA | AK029835.1 | - | 12 | 2233 | ||
| AK046518.1 | - | 12 | 1896 | |||
| AK122530.1 | - | 10 | 1382 | |||
| AK147386.1 | - | 10 | 1393 | |||
| AK172373.1 | - | 12 | 1868 | |||
| AK189788.1 | - | 2 | 51 | |||
| AK193535.1 | - | 2 | 51 | |||
| BC023048.1 | - | 3 | 375 | |||
| BC079562.1 | - | 5 | 494 | |||

Fam160b1
Genomic Map