siRNA Details
| Gene Symbol: 5430435G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145509.2 | NP_663484.1 | 6 | 2197 | ||
| GenBank mRNA | AK030688.1 | - | 6 | 2198 | ||
| AK079999.1 | - | 6 | 1769 | |||
| AK163610.1 | - | 1 | 648 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430435G22Rik| Gene Symbol: 5430435G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145509.2 | NP_663484.1 | 6 | 2197 | ||
| GenBank mRNA | AK030688.1 | - | 6 | 2198 | ||
| AK079999.1 | - | 6 | 1769 | |||
| AK163610.1 | - | 1 | 648 | |||