siRNA Details
| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145515.2 | NP_663490.2 | 18 | 3147 | ||
| GenBank mRNA | AF453686.1 | - | 18 | 3079 | ||
| AK047140.1 | - | 1 | 237 | |||
| AK136694.1 | - | 1 | 414 | |||
| AK173181.1 | - | 17 | 3000 | |||

Mark1
Genomic Map