siRNA Details
| Gene Symbol: Ormdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145517.4 | NP_663492.3 | 4 | 1069 | ||
| GenBank mRNA | AK050385.1 | - | 4 | 1054 | ||
| AK077562.1 | - | 5 | 1132 | |||
| BC012315.1 | - | 4 | 1024 | 3981845 | ||
| BC023695.1 | - | 4 | 1049 | 5344622 | ||
| BC023903.1 | - | 4 | 1012 | |||
| BC025572.1 | - | 4 | 1045 | 5323819 | ||
| EL956043.1 | - | 1 | 567 | |||

Ormdl1
Genomic Map