siRNA Details
| Gene Symbol: Gca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145523.3 | NP_663498.1 | 7 | 724 | ||
| GenBank mRNA | AB088388.1 | - | 7 | 618 | ||
| AF518325.1 | - | 7 | 708 | |||
| AK033740.1 | - | 7 | 757 | |||
| AK046347.1 | - | 7 | 766 | |||
| AK049317.1 | - | 7 | 725 | |||
| AK050540.1 | - | 7 | 766 | |||
| AK078915.1 | - | 7 | 725 | |||
| BC021450.1 | - | 7 | 718 | 5044040 | ||

Gca
Genomic Map