siRNA Details
| Gene Symbol: Fbxo21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145564.3 | NP_663539.1 | 9, 10 | 1360 | ||
| GenBank mRNA | AK049723.1 | - | 9, 10 | 1361 | ||
| BC021871.1 | - | 9, 10 | 1397 | 3709413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo21| Gene Symbol: Fbxo21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145564.3 | NP_663539.1 | 9, 10 | 1360 | ||
| GenBank mRNA | AK049723.1 | - | 9, 10 | 1361 | ||
| BC021871.1 | - | 9, 10 | 1397 | 3709413 | ||