siRNA Details
| Gene Symbol: Zfp772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145577.2 | NP_663552.2 | 2, 3 | 322 | ||
| GenBank mRNA | AK142307.1 | - | 2, 3 | 322 | ||
| BC023179.1 | - | 2, 3 | 278 | 4951074 | ||
| BC108353.1 | - | 2, 3 | 213 | 5067589 | ||

Zfp772
Genomic Map