siRNA Details
| Gene Symbol: BC017158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145590.2 | NP_663565.2 | 6 | 712 | ||
| GenBank mRNA | AK148236.1 | - | 6 | 703 | ||
| AK149645.1 | - | 6 | 691 | |||
| AK154566.1 | - | 6 | 678 | |||
| AK164488.1 | - | 6 | 692 | |||
| AK165287.1 | - | 6 | 691 | |||
| AK170611.1 | - | 6 | 691 | |||
| BC017158.1 | - | 6 | 694 | 4919869 | ||
| BC024080.1 | - | 5 | 322 | |||
| BC094040.1 | - | 6 | 676 | |||

BC017158
Genomic Map