siRNA Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145600.1 | NP_663575.1 | 6 | 425 | ||
| GenBank mRNA | AK029489.1 | - | 5 | 250 | ||
| AK037748.1 | - | 2 | 707 | |||
| AK050534.1 | - | 6 | 435 | |||
| AK086617.1 | - | 6 | 459 | |||
| BC008086.1 | - | 6 | 425 | 3586161 | ||
| BC030675.1 | - | 6 | 423 | 1068780 | ||
| BC037025.1 | - | 2 | 1782 | |||
| BC083081.1 | - | 6 | 459 | 6510396 | ||

Zfp330
Genomic Map