siRNA Details
| Gene Symbol: Zfp810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145612.2 | NP_663587.1 | 6, 7 | 606 | ||
| GenBank mRNA | AK032556.1 | - | 6, 7 | 646 | ||
| BC005471.1 | - | 6, 7 | 606 | 3496918 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp810| Gene Symbol: Zfp810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145612.2 | NP_663587.1 | 6, 7 | 606 | ||
| GenBank mRNA | AK032556.1 | - | 6, 7 | 646 | ||
| BC005471.1 | - | 6, 7 | 606 | 3496918 | ||