siRNA Details
| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167775.1 | NP_001161247.1 | 3 | 781 | ||
| NM_001167776.1 | NP_001161248.1 | 3 | 494 | |||
| NM_145627.2 | NP_663602.1 | 3 | 494 | |||
| GenBank mRNA | AK041871.1 | - | 3 | 469 | ||
| AK089105.1 | - | 3 | 305 | |||
| AK122208.1 | - | 3 | 412 | |||
| AK145991.1 | - | 3 | 494 | |||
| AK153736.1 | - | 3 | 782 | |||
| AK167666.1 | - | 3 | 472 | |||
| BC004674.1 | - | 2 | 179 | 3500403 | ||
| BC093519.1 | - | 3 | 426 | |||
| Gene Symbol: Gm15455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161816.1 | NP_001155288.1 | 1 | 311 | ||
| GenBank mRNA | AK139531.1 | - | 1060 | |||
| BC157984.1 | - | 1 | 235 | |||
| BC157993.1 | - | 1 | 235 | |||

Rbm10
Genomic Map