siRNA Details
| Gene Symbol: BLVRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000713.2 | NP_000704.1 | 3 | 435 | ||
| GenBank mRNA | AK312137.1 | - | 3 | 362 | ||
| BC109371.1 | - | 3 | 352 | 5229026 | ||
| BG340491.1 | - | 3 | 435 | |||
| BU528444.1 | - | 1 | 30 | |||
| D26308.1 | - | 3 | 383 | |||
| D32143.1 | - | 3 | 414 | |||
| EU794593.1 | - | 3 | 435 | |||

BLVRB
Genomic Map