siRNA Details
| Gene Symbol: 9430015G10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145557.3 | NP_663532.2 | 10 | 675 | ||
| NM_177205.1 | NP_796179.1 | 10 | 825 | |||
| GenBank mRNA | AK029276.1 | - | 8 | 1101 | ||
| AK034620.1 | - | 9 | 539 | |||
| BC004010.1 | - | 10 | 695 | 3493145 | ||
| CA945382.1 | - | 5 | 226 | |||

9430015G10Rik
Genomic Map