siRNA Details
Gene Symbol: Eva1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145570.2 | NP_663545.2 | 3 | 819 | ||
GenBank mRNA | BC014699.1 | - | 3 | 752 | 4221746 | |
BC027150.1 | - | 3 | 774 | 4950341 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eva1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145570.2 | NP_663545.2 | 3 | 819 | ||
GenBank mRNA | BC014699.1 | - | 3 | 752 | 4221746 | |
BC027150.1 | - | 3 | 774 | 4950341 |