siRNA Details
| Gene Symbol: Tmem184c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145599.4 | NP_663574.3 | 2, 3 | 690 | ||
| GenBank mRNA | AK033404.1 | - | 2, 3 | 659 | ||
| AK164182.1 | - | 2, 3 | 658 | |||
| BC004056.1 | - | 2, 3 | 648 | 3589819 | ||
| CN532034.1 | - | 2, 3 | 690 | |||

Tmem184c
Genomic Map