siRNA Details
| Gene Symbol: Eif4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145625.3 | NP_663600.2 | 10 | 1442 | ||
| GenBank mRNA | AK045250.1 | - | 10 | 1286 | ||
| AK077423.1 | - | 10 | 1293 | |||
| AK148012.1 | - | 10 | 1292 | |||
| AK161693.1 | - | 10 | 1275 | |||
| AK163266.1 | - | 10 | 1293 | |||
| AK166363.1 | - | 10 | 1294 | |||
| AK168350.1 | - | 10 | 1288 | |||
| AK168425.1 | - | 10 | 1288 | |||
| AK170256.1 | - | 10 | 1293 | |||
| AK170354.1 | - | 10 | 1275 | |||
| AK178737.1 | - | 1 | 64 | |||
| AK180578.1 | - | 1 | 72 | |||
| AK206900.1 | - | 1 | 64 | |||
| AK214347.1 | - | 1 | 64 | |||
| AK219884.1 | - | 1 | 20 | |||
| BC007171.1 | - | 9 | 1241 | 3492114 | ||
| BC036293.1 | - | 10 | 1278 | |||

Eif4b
Genomic Map