siRNA Details
| Gene Symbol: Zfp772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145577.2 | NP_663552.2 | 5 | 708 | ||
| GenBank mRNA | AK142307.1 | - | 5 | 708 | ||
| BC023179.1 | - | 5 | 664 | 4951074 | ||
| BC108353.1 | - | 5 | 599 | 5067589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp772| Gene Symbol: Zfp772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145577.2 | NP_663552.2 | 5 | 708 | ||
| GenBank mRNA | AK142307.1 | - | 5 | 708 | ||
| BC023179.1 | - | 5 | 664 | 4951074 | ||
| BC108353.1 | - | 5 | 599 | 5067589 | ||