siRNA Details
| Gene Symbol: BC017158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145590.2 | NP_663565.2 | 11, 12 | 1277 | ||
| GenBank mRNA | AK148236.1 | - | 11, 12 | 1268 | ||
| AK149645.1 | - | 11, 12 | 1256 | |||
| AK164488.1 | - | 11, 12 | 1255 | |||
| AK165287.1 | - | 9, 10 | 2720 | |||
| AK170611.1 | - | 10, 11 | 1340 | |||
| BC017158.1 | - | 11, 12 | 1259 | 4919869 | ||
| BC024080.1 | - | 10, 11 | 887 | |||
| BC061499.1 | - | 1, 2 | 1203 | |||
| BC094040.1 | - | 11, 12 | 1261 | |||

BC017158
Genomic Map