siRNA Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293797.1 | NP_001280726.1 | 8 | 830 | ||
| NM_145597.4 | NP_663572.1 | 9 | 934 | |||
| GenBank mRNA | AK032711.1 | - | 5 | 450 | ||
| AK049979.1 | - | 9 | 933 | |||
| AK082916.1 | - | 8 | 831 | |||
| AK088569.1 | - | 8 | 820 | |||
| AK147962.1 | - | 9 | 927 | |||
| BC021367.1 | - | 9 | 934 | 5030434 | ||

Tmem161a
Genomic Map