siRNA Details
| Gene Symbol: Zfp330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145600.1 | NP_663575.1 | 8, 9 | 660 | ||
| GenBank mRNA | AK029489.1 | - | 7, 8 | 485 | ||
| AK050534.1 | - | 8, 9 | 670 | |||
| AK086617.1 | - | 8, 9 | 694 | |||
| BC008086.1 | - | 8, 9 | 660 | 3586161 | ||
| BC030675.1 | - | 8, 9 | 658 | 1068780 | ||
| BC037025.1 | - | 4, 5 | 2017 | |||
| BC083081.1 | - | 8, 9 | 694 | 6510396 | ||

Zfp330
Genomic Map