siRNA Details
| Gene Symbol: DHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000791.3 | NP_000782.1 | 6 | 1694 | ||
| NM_001290354.1 | NP_001277283.1 | 5 | 1644 | |||
| NM_001290357.1 | NP_001277286.1 | 5 | 1578 | |||
| NR_110936.1 | 4 | 1517 | ||||
| GenBank mRNA | AK293146.1 | - | 5 | 1222 | ||
| AK297302.1 | - | 5 | 1149 | |||
| BC000192.1 | - | 6 | 1257 | 3352539 | ||

DHFR
Genomic Map