siRNA Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145823.2 | NP_665822.1 | 3 | 1045 | ||
| GenBank mRNA | AB077281.1 | - | 3 | 1026 | ||
| AB077282.1 | - | 3 | 323 | |||
| AK003844.1 | - | 2 | 293 | |||
| AK021206.1 | - | 3 | 258 | |||
| AK153625.1 | - | 3 | 1046 | |||
| AK171340.1 | - | 3 | 1072 | |||
| AK171918.1 | - | 3 | 1042 | |||
| AK172137.1 | - | 3 | 1046 | |||
| BC082333.1 | - | 3 | 877 | |||
| BC108351.1 | - | 3 | 905 | 6312193 | ||

Pitpnc1
Genomic Map