siRNA Details
| Gene Symbol: Cetn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145825.2 | NP_665824.1 | 2 | 188 | ||
| GenBank mRNA | AF362367.1 | - | 2 | 188 | ||
| AK078275.1 | - | 1 | 2103 | |||
| AK090064.1 | - | 175 | ||||
| AK131808.1 | - | 2 | 176 | |||
| AK166515.1 | - | 1 | 104 | |||
| BC038154.1 | - | 1 | 79 | |||
| BC060991.1 | - | 2 | 140 | 6742836 | ||
| BC087905.1 | - | 2 | 166 | 6707456 | ||

Cetn4
Genomic Map