siRNA Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145828.3 | NP_665827.2 | 3 | 853 | ||
| GenBank mRNA | AK083482.1 | - | 3 | 822 | ||
| AK086804.1 | - | 3 | 785 | |||
| AK150521.1 | - | 3 | 806 | |||
| AK153332.1 | - | 3 | 806 | |||
| AK158056.1 | - | 3 | 853 | |||
| BC034082.1 | - | 2 | 372 | 4239646 | ||

Xylt2
Genomic Map