siRNA Details
| Gene Symbol: Vmn1r192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145845.1 | NP_665844.1 | 1 | 192 | ||
| GenBank mRNA | BC138395.1 | - | 1 | 285 | 8861415 | |
| BC138400.1 | - | 1 | 284 | 8861420 | ||
| Gene Symbol: Vmn1r194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080972.1 | NP_001074441.1 | 1 | 192 | ||

Vmn1r192
Genomic Map